siRNA Details
| Gene Symbol: Plod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142915.1 | NP_001136387.1 | 3 | 465 | ||
| NM_175869.3 | NP_787065.2 | 3 | 465 | |||
| GenBank mRNA | AJ430860.1 | - | 3 | 527 | ||
| AJ430861.1 | - | 3 | 527 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plod2| Gene Symbol: Plod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142915.1 | NP_001136387.1 | 3 | 465 | ||
| NM_175869.3 | NP_787065.2 | 3 | 465 | |||
| GenBank mRNA | AJ430860.1 | - | 3 | 527 | ||
| AJ430861.1 | - | 3 | 527 | |||