siRNA Details
| Gene Symbol: Plod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142915.1 | NP_001136387.1 | 17 | 2072 | ||
| NM_175869.3 | NP_787065.2 | 18 | 2135 | |||
| GenBank mRNA | AJ430860.1 | - | 18 | 2197 | ||
| AJ430861.1 | - | 17 | 2134 | |||

Plod2
Genomic Map