siRNA Details
| Gene Symbol: Mrpl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106852.1 | NP_001100322.1 | 5 | 689 | ||
| GenBank mRNA | FQ233139.1 | - | 1 | 43 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl3| Gene Symbol: Mrpl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106852.1 | NP_001100322.1 | 5 | 689 | ||
| GenBank mRNA | FQ233139.1 | - | 1 | 43 | ||