siRNA Details
| Gene Symbol: Cacna2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175592.2 | NP_783182.1 | 16, 17 | 1560 | ||
| GenBank mRNA | AF486277.1 | - | 16, 17 | 1560 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacna2d2| Gene Symbol: Cacna2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175592.2 | NP_783182.1 | 16, 17 | 1560 | ||
| GenBank mRNA | AF486277.1 | - | 16, 17 | 1560 | ||