siRNA Details
| Gene Symbol: Impdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199099.2 | NP_954530.1 | 12 | 1436 | ||
| XM_006243751.2 | XP_006243813.1 | 13 | 1549 | |||
| GenBank mRNA | BC060585.1 | - | 12 | 1437 | 6891123 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Impdh2| Gene Symbol: Impdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199099.2 | NP_954530.1 | 12 | 1436 | ||
| XM_006243751.2 | XP_006243813.1 | 13 | 1549 | |||
| GenBank mRNA | BC060585.1 | - | 12 | 1437 | 6891123 | |