siRNA Details
Gene Symbol: Nckipsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106857.1 | NP_001100327.1 | 2 | 282 | ||
XM_006243753.2 | XP_006243815.1 | 2 | 287 | |||
XM_006243754.2 | XP_006243816.1 | 2 | 632 | |||
XM_008766508.1 | XP_008764730.1 | 2 | 2400 | |||
XM_008766509.1 | XP_008764731.1 | 3 | 426 |
