siRNA Details
| Gene Symbol: Col7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106858.1 | NP_001100328.1 | 15 | 1975 | ||
| XM_006243759.2 | XP_006243821.1 | 16 | 2030 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col7a1| Gene Symbol: Col7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106858.1 | NP_001100328.1 | 15 | 1975 | ||
| XM_006243759.2 | XP_006243821.1 | 16 | 2030 | |||