siRNA Details
| Gene Symbol: Ctdspl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106865.1 | NP_001100335.1 | 7 | 712 | ||
| XM_008766675.1 | XP_008764897.1 | 4 | 3578 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctdspl| Gene Symbol: Ctdspl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106865.1 | NP_001100335.1 | 7 | 712 | ||
| XM_008766675.1 | XP_008764897.1 | 4 | 3578 | |||