siRNA Details
| Gene Symbol: Myd88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198130.1 | NP_937763.1 | 1 | 382 | ||
| XM_006244087.2 | XP_006244149.1 | 2 | 729 | |||
| GenBank mRNA | AY191270.1 | - | 1 | 382 | ||
| BC097266.1 | - | 1 | 322 | 7459961 | ||

Myd88
Genomic Map