siRNA Details
| Gene Symbol: Ttc21a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750604.3 | XP_003750652.1 | 4 | 512 | ||
| XM_006244150.2 | XP_006244212.1 | 4 | 512 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc21a| Gene Symbol: Ttc21a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750604.3 | XP_003750652.1 | 4 | 512 | ||
| XM_006244150.2 | XP_006244212.1 | 4 | 512 | |||