siRNA Details
| Gene Symbol: Zfp445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191802.1 | NP_001178731.1 | 7 | 1794 | ||
| XM_006244181.1 | XP_006244243.1 | 7 | 1868 | |||
| XM_006244183.2 | XP_006244245.1 | 7 | 1795 | |||
| GenBank mRNA | FQ226616.1 | - | 6 | 1541 | ||

Zfp445
Genomic Map