siRNA Details
| Gene Symbol: Cdcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106869.1 | NP_001100339.1 | 9 | 2225 | ||
| XM_006244189.2 | XP_006244251.1 | 9 | 2216 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdcp1| Gene Symbol: Cdcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106869.1 | NP_001100339.1 | 9 | 2225 | ||
| XM_006244189.2 | XP_006244251.1 | 9 | 2216 | |||