siRNA Details
| Gene Symbol: Fyco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106870.1 | NP_001100340.1 | 9 | 1794 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fyco1| Gene Symbol: Fyco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106870.1 | NP_001100340.1 | 9 | 1794 | ||