siRNA Details
| Gene Symbol: Acer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106875.1 | NP_001100345.1 | 2 | 229 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acer1| Gene Symbol: Acer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106875.1 | NP_001100345.1 | 2 | 229 | ||