siRNA Details
| Gene Symbol: Acsbg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080096.1 | NP_001073565.1 | 3, 4 | 548 | ||
| XM_006244311.2 | XP_006244373.1 | 3, 4 | 1057 | |||
| XM_006244312.2 | XP_006244374.1 | 3, 4 | 1059 | |||
| GenBank mRNA | BC129110.1 | - | 3, 4 | 548 | 7111052 | |

Acsbg2
Genomic Map