siRNA Details
| Gene Symbol: Yipf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007801.2 | NP_001007802.1 | 6 | 738 | ||
| GenBank mRNA | AY387096.1 | - | 6 | 633 | ||
| BC087102.1 | - | 6 | 738 | 7192818 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yipf3| Gene Symbol: Yipf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007801.2 | NP_001007802.1 | 6 | 738 | ||
| GenBank mRNA | AY387096.1 | - | 6 | 633 | ||
| BC087102.1 | - | 6 | 738 | 7192818 | ||