siRNA Details
| Gene Symbol: Polr1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008330.1 | NP_001008331.1 | 9 | 1065 | ||
| GenBank mRNA | AY325219.1 | - | 7 | 791 | ||
| BC086597.1 | - | 9 | 1065 | 7312277 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Polr1c| Gene Symbol: Polr1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008330.1 | NP_001008331.1 | 9 | 1065 | ||
| GenBank mRNA | AY325219.1 | - | 7 | 791 | ||
| BC086597.1 | - | 9 | 1065 | 7312277 | ||