siRNA Details
Gene Symbol: Aars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106891.1 | NP_001100361.1 | 9, 10 | 1300 | ||
XM_006244513.2 | XP_006244575.1 | 9, 10 | 2290 | |||
XM_006244515.2 | XP_006244577.1 | 9, 10 | 2286 | |||
GenBank mRNA | BC166719.1 | - | 9, 10 | 1274 |
