siRNA Details
| Gene Symbol: Enpp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106892.1 | NP_001100362.1 | 4 | 1324 | ||
| XM_006244598.2 | XP_006244660.1 | 5 | 2720 | |||
| XM_006244600.2 | XP_006244662.1 | 4 | 2605 | |||
| XM_006244601.2 | XP_006244663.1 | 6 | 2369 | |||
| XM_006244602.2 | XP_006244664.1 | 4 | 1306 | |||
| XM_008766852.1 | XP_008765074.1 | 5 | 1420 | |||
| XM_008766853.1 | XP_008765075.1 | 4 | 1214 | |||

Enpp4
Genomic Map