siRNA Details
| Gene Symbol: Pla2g7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009353.1 | NP_001009353.1 | 8, 9 | 995 | ||
| XM_006244606.2 | XP_006244668.1 | 8, 9 | 1112 | |||
| XM_008766856.1 | XP_008765078.1 | 7, 8 | 2755 | |||
| GenBank mRNA | BC088457.1 | - | 8, 9 | 995 | 7127300 | |

Pla2g7
Genomic Map