siRNA Details
| Gene Symbol: Tfap2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106895.1 | NP_001100365.1 | 7 | 905 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tfap2d| Gene Symbol: Tfap2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106895.1 | NP_001100365.1 | 7 | 905 | ||