siRNA Details
| Gene Symbol: Efhc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122947.2 | NP_001116419.2 | 9 | 1554 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Efhc1| Gene Symbol: Efhc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122947.2 | NP_001116419.2 | 9 | 1554 | ||