siRNA Details
| Gene Symbol: Prim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024762.1 | NP_001019933.1 | 4 | 355 | ||
| XM_006244686.2 | XP_006244748.1 | 2 | 184 | |||
| XM_006244689.2 | XP_006244751.1 | 2 | 135 | |||
| XM_006244690.2 | XP_006244752.1 | 2 | 201 | |||
| XM_006244691.1 | XP_006244753.1 | 2 | 202 | |||
| GenBank mRNA | AJ011607.1 | - | 1 | 39 | ||
| BC097454.1 | - | 4 | 355 | 7456950 | ||

Prim2
Genomic Map