siRNA Details
| Gene Symbol: Mrps9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100549.1 | NP_001094019.1 | 2 | 226 | ||
| XM_008767019.1 | XP_008765241.1 | 2 | 237 | |||
| GenBank mRNA | BC088242.1 | - | 1 | 38 | ||
| BC158704.1 | - | 2 | 194 | 9030006 | ||
| CV106852.1 | - | 2 | 226 | |||

Mrps9
Genomic Map