siRNA Details
| Gene Symbol: Hibch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013112.1 | NP_001013130.1 | 7 | 551 | ||
| XM_006244895.1 | XP_006244957.1 | 7 | 637 | |||
| GenBank mRNA | BC083737.1 | - | 7 | 551 | 7191666 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hibch| Gene Symbol: Hibch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013112.1 | NP_001013130.1 | 7 | 551 | ||
| XM_006244895.1 | XP_006244957.1 | 7 | 637 | |||
| GenBank mRNA | BC083737.1 | - | 7 | 551 | 7191666 | |