siRNA Details
| Gene Symbol: Orc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012003.1 | NP_001012003.1 | 11 | 1086 | ||
| XM_006244956.2 | XP_006245018.1 | 12 | 1372 | |||
| XM_008767081.1 | XP_008765303.1 | 9 | 2029 | |||
| GenBank mRNA | AB164704.1 | - | 11 | 1086 | ||
| BC129076.1 | - | 10 | 1695 | |||

Orc2
Genomic Map