siRNA Details
| Gene Symbol: Ppil3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175707.3 | NP_783638.1 | 6 | 636 | ||
| XM_006244964.2 | XP_006245026.1 | 6 | 706 | |||
| GenBank mRNA | BC087645.1 | - | 6 | 636 | 7311008 | |
| FQ224392.1 | - | 6 | 433 | |||

Ppil3
Genomic Map