siRNA Details
| Gene Symbol: LOC100360552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271185.1 | NP_001258114.1 | 1 | 1373 | ||
More Information
Other Species Targeted
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100360552| Gene Symbol: LOC100360552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271185.1 | NP_001258114.1 | 1 | 1373 | ||
Set Membership: |
|