siRNA Details
Gene Symbol: Sumo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009672.1 | NP_001009672.1 | 3, 4 | 207 | ||
GenBank mRNA | BC088322.1 | - | 3, 4 | 207 | 7325996 | |
FQ211636.1 | - | 3, 4 | 244 | |||
FQ214389.1 | - | 3, 4 | 294 | |||
FQ214927.1 | - | 3, 4 | 277 | |||
FQ215001.1 | - | 3, 4 | 287 | |||
FQ215643.1 | - | 3, 4 | 252 | |||
FQ224870.1 | - | 3, 4 | 276 | |||
FQ234204.1 | - | 3, 4 | 279 |
