siRNA Details
Gene Symbol: Carf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106915.1 | NP_001100385.1 | 17 | 2132 | ||
XM_006244969.1 | XP_006245031.1 | 14 | 1994 | |||
XM_006244970.2 | XP_006245032.1 | 15 | 2280 | |||
XM_006244971.1 | XP_006245033.1 | 13 | 1835 | |||
XM_006244973.2 | XP_006245035.1 | 12 | 1556 | |||
XM_006244974.2 | XP_006245036.1 | 13 | 2520 |
