siRNA Details
| Gene Symbol: Pard3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191808.1 | NP_001178737.1 | 23 | 3315 | ||
| XM_008767088.1 | XP_008765310.1 | 22 | 3141 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pard3b| Gene Symbol: Pard3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191808.1 | NP_001178737.1 | 23 | 3315 | ||
| XM_008767088.1 | XP_008765310.1 | 22 | 3141 | |||