siRNA Details
| Gene Symbol: Bcs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007666.1 | NP_001007667.1 | 2 | 338 | ||
| GenBank mRNA | BC083660.1 | - | 2 | 338 | 7135331 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcs1l| Gene Symbol: Bcs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007666.1 | NP_001007667.1 | 2 | 338 | ||
| GenBank mRNA | BC083660.1 | - | 2 | 338 | 7135331 | |