siRNA Details
| Gene Symbol: Armc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109663.1 | NP_001103133.1 | 20 | 1901 | ||
| GenBank mRNA | BC128749.1 | - | 20 | 1870 | 7099560 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Armc9| Gene Symbol: Armc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109663.1 | NP_001103133.1 | 20 | 1901 | ||
| GenBank mRNA | BC128749.1 | - | 20 | 1870 | 7099560 | |