siRNA Details
| Gene Symbol: Pask | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009362.1 | NP_001009362.1 | 7 | 1283 | ||
| GenBank mRNA | BC087047.1 | - | 7 | 1283 | 7112450 | |
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Pask
Pask| Gene Symbol: Pask | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009362.1 | NP_001009362.1 | 7 | 1283 | ||
| GenBank mRNA | BC087047.1 | - | 7 | 1283 | 7112450 | |
 Back To Top
 Back To Top