siRNA Details
| Gene Symbol: Ppp1r7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009825.1 | NP_001009825.1 | 3, 4 | 246 | ||
| XM_008767362.1 | XP_008765584.1 | 3, 4 | 1192 | |||
| GenBank mRNA | BC088868.1 | - | 3, 4 | 246 | 7321115 | |
| FQ233576.1 | - | 260 | ||||

Ppp1r7
Genomic Map