siRNA Details
| Gene Symbol: Fbxo11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181631.2 | NP_853662.1 | 5 | 515 | ||
| XM_008764477.1 | XP_008762699.1 | 5 | 2289 | |||
| GenBank mRNA | AY274810.1 | - | 5 | 515 | ||
| Gene Symbol: LOC103689957 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239732.2 | XP_006239794.1 | 5 | 2299 | ||
| XM_006239733.2 | XP_006239795.1 | 5 | 2296 | |||

Fbxo11
Genomic Map