siRNA Details
| Gene Symbol: Fbxo11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181631.2 | NP_853662.1 | 14, 15 | 1689 | ||
| XM_008764477.1 | XP_008762699.1 | 14, 15 | 3463 | |||
| GenBank mRNA | AY274810.1 | - | 14, 15 | 1689 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo11| Gene Symbol: Fbxo11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181631.2 | NP_853662.1 | 14, 15 | 1689 | ||
| XM_008764477.1 | XP_008762699.1 | 14, 15 | 3463 | |||
| GenBank mRNA | AY274810.1 | - | 14, 15 | 1689 | ||