siRNA Details
| Gene Symbol: Copg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106929.2 | NP_001100399.1 | 9, 10 | 841 | ||
| GenBank mRNA | FM091200.1 | - | 3, 4 | 272 | ||
| FN805880.1 | - | 7, 8 | 621 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Copg2| Gene Symbol: Copg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106929.2 | NP_001100399.1 | 9, 10 | 841 | ||
| GenBank mRNA | FM091200.1 | - | 3, 4 | 272 | ||
| FN805880.1 | - | 7, 8 | 621 | |||