siRNA Details
| Gene Symbol: Copg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106929.2 | NP_001100399.1 | 11, 12 | 1035 | ||
| GenBank mRNA | FM091200.1 | - | 5, 6 | 466 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Copg2| Gene Symbol: Copg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106929.2 | NP_001100399.1 | 11, 12 | 1035 | ||
| GenBank mRNA | FM091200.1 | - | 5, 6 | 466 | ||