siRNA Details
Gene Symbol: Tert | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053423.1 | NP_445875.1 | 2 | 687 | ||
XM_008758683.1 | XP_008756905.1 | 2 | 1934 | |||
XM_008758684.1 | XP_008756906.1 | 2 | 1934 | |||
GenBank mRNA | AY539717.1 | - | 2 | 687 | ||
AY539718.1 | - | 2 | 687 | |||
AY539719.1 | - | 2 | 687 | |||
AY539720.1 | - | 2 | 687 |
