siRNA Details
| Gene Symbol: LOC103690878 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773369.1 | XP_008771591.1 | 1 | 799 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103690878| Gene Symbol: LOC103690878 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773369.1 | XP_008771591.1 | 1 | 799 | ||