siRNA Details
| Gene Symbol: Tbx22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106937.1 | NP_001100407.1 | 3, 4 | 396 | ||
| XM_006257159.2 | XP_006257221.1 | 4, 5 | 665 | |||
| XM_006257160.2 | XP_006257222.1 | 4, 5 | 515 | |||
| XM_006257161.2 | XP_006257223.1 | 4, 5 | 662 | |||
| XM_006257162.2 | XP_006257224.1 | 3, 4 | 705 | |||
| XM_008773339.1 | XP_008771561.1 | 3, 4 | 380 | |||

Tbx22
Genomic Map