siRNA Details
| Gene Symbol: Gpr174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106938.1 | NP_001100408.1 | 2 | 1070 | ||
| XM_006257163.2 | XP_006257225.1 | 4 | 1291 | |||
| XM_006257164.2 | XP_006257226.1 | 4 | 1287 | |||
| XM_008773340.1 | XP_008771562.1 | 4 | 1302 | |||
More Information
Set Membership: |
|

Gpr174
Genomic Map