siRNA Details
| Gene Symbol: RGD1560455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106939.1 | NP_001100409.1 | 2 | 1266 | ||
| XM_006257166.2 | XP_006257228.1 | 2 | 1770 | |||
| XM_006257168.2 | XP_006257230.1 | 2 | 1193 | |||
| XM_006257169.2 | XP_006257231.1 | 2 | 1376 | |||

RGD1560455
Genomic Map