siRNA Details
| Gene Symbol: Lpar4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106940.1 | NP_001100410.1 | 3 | 1455 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lpar4| Gene Symbol: Lpar4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106940.1 | NP_001100410.1 | 3 | 1455 | ||