siRNA Details
| Gene Symbol: Magebl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128493.1 | NP_001121965.1 | 3 | 1061 | ||
| XM_006257011.2 | XP_006257073.1 | 3 | 1057 | |||
| GenBank mRNA | BC158628.1 | - | 3 | 1061 | 7455432 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Magebl1| Gene Symbol: Magebl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128493.1 | NP_001121965.1 | 3 | 1061 | ||
| XM_006257011.2 | XP_006257073.1 | 3 | 1057 | |||
| GenBank mRNA | BC158628.1 | - | 3 | 1061 | 7455432 | |