siRNA Details
Gene Symbol: Magebl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128493.1 | NP_001121965.1 | 3 | 325 | ||
XM_006257011.2 | XP_006257073.1 | 3 | 321 | |||
GenBank mRNA | BC158628.1 | - | 3 | 325 | 7455432 |
