siRNA Details
| Gene Symbol: Mbnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106949.1 | NP_001100419.1 | 1 | 198 | ||
| XM_008773556.1 | XP_008771778.1 | 1 | 1677 | |||
| XM_008773557.1 | XP_008771779.1 | 1 | 800 | |||
| XM_008773558.1 | XP_008771780.1 | 1 | 797 | |||
| XM_008773559.1 | XP_008771781.1 | 1 | 800 | |||
| XM_008773560.1 | XP_008771782.1 | 1 | 803 | |||
| XM_008773561.1 | XP_008771783.1 | 1 | 800 | |||

Mbnl3
Genomic Map