siRNA Details
| Gene Symbol: Atp6ap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007091.1 | NP_001007092.1 | 5 | 605 | ||
| GenBank mRNA | AB188298.1 | - | 5 | 605 | ||
| BC079339.1 | - | 605 | 7106562 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp6ap2| Gene Symbol: Atp6ap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007091.1 | NP_001007092.1 | 5 | 605 | ||
| GenBank mRNA | AB188298.1 | - | 5 | 605 | ||
| BC079339.1 | - | 605 | 7106562 | |||