siRNA Details
| Gene Symbol: Acot9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013960.1 | NP_001013982.1 | 9, 10 | 790 | ||
| GenBank mRNA | BC085822.1 | - | 9, 10 | 790 | 7126662 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acot9| Gene Symbol: Acot9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013960.1 | NP_001013982.1 | 9, 10 | 790 | ||
| GenBank mRNA | BC085822.1 | - | 9, 10 | 790 | 7126662 | |